MilliporeSigma
  • Modular synthesis of indoles from imines and o-dihaloarenes or o-chlorosulfonates by a Pd-catalyzed cascade process.
Modular synthesis of indoles from imines and o-dihaloarenes or o-chlorosulfonates by a Pd-catalyzed cascade process.
Journal of the American Chemical Society (2009-02-28)
José Barluenga, Agustín Jiménez-Aquino, Fernando Aznar, Carlos Valdés
ABSTRACT
A detailed study of the scope of a new Pd-catalyzed synthesis of indoles from 1,2-dihaloarenes and o-halobenzene sulfonates and imines is described. The cascade reaction comprises an imine alpha-arylation followed by an intramolecular C-N bond-forming reaction promoted by the same Pd catalyst. The reaction with 1,2-dibromobenzene shows wide scope and allows the introduction of aryl, alkyl, and vinyl substituents at different positions of the five-membered ring of the indole. The regioselective synthesis of indoles substituted in the six-membered ring can be carried out by employing o-dihalobenzene derivatives with two different halogens, taking advantage of the different reactivities of I, Br, and Cl in oxidative addition reactions. This paper also introduces a method for the efficient cleavage of the N-t-butyl group, thus allowing for the preparation of N-H indoles through the same methodology. Finally, the reaction with o-halosulfonates has been studied. The best substrates are o-chlorononaflates, which lead to indoles in very high yield. The reaction is particularly appropriate for the synthesis of the challenging 6-substituted indoles. In view of the availability of o-chlorophenols, which are direct precursors of the chlorononaflates, this reaction represents an efficient entry into indoles substituted in the six-membered ring. The concept is illustrated by the preparation of a 4,6-disubstituted indole from naturally occurring anethole.
Sigma-Aldrich
Chlorosulfonic acid, 99%