MilliporeSigma
  • Chiral self-discrimination of the enantiomers of alpha-phenylethylamine derivatives in proton NMR.
Chiral self-discrimination of the enantiomers of alpha-phenylethylamine derivatives in proton NMR.
Magnetic resonance in chemistry : MRC (2009-02-19)
Shao-Hua Huang, Zheng-Wu Bai, Ji-Wen Feng
ABSTRACT
Two types of chiral analytes, the urea and amide derivatives of alpha-phenylethylamine, were prepared. The effect of inter-molecular hydrogen-bonding interaction on self-discrimination of the enantiomers of analytes has been investigated using high-resolution (1)H NMR. It was found that the urea derivatives with double-hydrogen-bonding interaction exhibit not only the stronger hydrogen-bonding interaction but also better self-recognition abilities than the amide derivatives (except for one bearing two NO(2) groups). The present results suggest that double-hydrogen-bonding interaction promotes the self-discrimination ability of the chiral compounds.
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamine, 98%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Sigma-Aldrich
α-Methylbenzylamine, 99%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, produced by BASF, ChiPros®, ≥99.0%
Supelco
(R)-(+)-α-Methylbenzylamine, for chiral derivatization, LiChropur, ≥99.0%