MilliporeSigma
  • Analysis of impurities in streptomycin and dihydrostreptomycin by hydrophilic interaction chromatography/electrospray ionization quadrupole ion trap/time-of-flight mass spectrometry.
Analysis of impurities in streptomycin and dihydrostreptomycin by hydrophilic interaction chromatography/electrospray ionization quadrupole ion trap/time-of-flight mass spectrometry.
Rapid communications in mass spectrometry : RCM (2009-02-19)
Shin-ichi Kawano
ABSTRACT
Impurities in streptomycin (STR) and dihydrostreptomycin (DHS) were investigated by hydrophilic interaction chromatography/electrospray ionization quadrupole ion trap/time-of-flight mass spectrometry (HILIC/ESI-QIT/TOFMS). Samples were separated on a fused-core silica column (100 mmx2.1 mm i.d., particle size: 2.7 microm) with isocratic elution using 200 mM ammonium formate buffer (pH 4.5) and acetonitrile as mobile phase. Constant neutral loss survey in accurate mass measurement was carried out by QIT/TOFMS. Formulae, chemical structures of impurities in an STR sample were suggested with supporting results on the probable pathways of STR biosynthesis by Streptomyces griseus.
Product Description
Dihydrostreptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dihydrostreptomycin sesquisulfate
Sigma-Aldrich
Dihydrostreptomycin sesquisulfate, BioReagent, suitable for cell culture, ≥95.0% (HPLC)