MilliporeSigma
  • Purification and characterization of a thermostable laccase with unique oxidative characteristics from Trametes hirsuta.
Purification and characterization of a thermostable laccase with unique oxidative characteristics from Trametes hirsuta.
Biotechnology letters (2009-02-18)
Zhang Haibo, Zhang Yinglong, Huang Feng, Gao Peiji, Chen Jiachuan
ABSTRACT
A laccase was purified from Trametes hirsuta. This laccase was classified as a "white" or "yellow" laccase. pH 2.4 was optimal for the oxidation of ABTS and pH 2.5 for DMP. DMP oxidation was optimal at 85 degrees C. The half-life of this laccase was 70 min at 75 degrees C, and 5 h at 65 degrees C. Non-phenolic dyes, such as Methyl Red, were oxidized by purified laccase without mediators. The enzyme was not inhibited by Cu(2+), Mn(2+), or EDTA. These are atypical laccase characteristics that make it a good candidate for theoretical and applied research.
Product Description
Millipore
Methyl Red solution, suitable for microbiology
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, indicator (pH 4.2-6.2)
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %