MilliporeSigma
  • In vitro remineralization of enamel by polymeric amorphous calcium phosphate composite: quantitative microradiographic study.
In vitro remineralization of enamel by polymeric amorphous calcium phosphate composite: quantitative microradiographic study.
Dental materials : official publication of the Academy of Dental Materials (2009-02-14)
S E Langhorst, J N R O'Donnell, D Skrtic
ABSTRACT
This study explores the efficacy of an experimental orthodontic amorphous calcium phosphate (ACP) composite to remineralize in vitro subsurface enamel lesions microradiographically similar to those seen in early caries. Lesions were artificially created in extracted human molars. Single tooth sections a minimum of 120microm thick were cut and individually placed in holders exposing only the carious enamel surface. The exposed surfaces were either left untreated (control) or coated with a 1mm thick layer of the experimental ACP composite (mass fraction 40% zirconia-hybridized ACP and 60% photo-activated resin), or a commercial fluoride-releasing orthodontic cement. The composite-coated sections were then photo-cured and microradiographic images were taken of all three groups of specimens before the treatment. Specimens were then cyclically immersed in demineralizing and remineralizing solutions for 1 month at 37 degrees C to simulate the pH changes occurring in the oral environment. Microradiographs of all specimens were taken before and after treatment. Quantitative digital image analysis of matched areas from the contact microradiographs taken before and after treatment indicated higher mineral recovery with ACP composites compared to the commercial orthodontic F-releasing cement (14.4% vs. 4.3%, respectively), while the control specimens showed an average of 55.4% further demineralization. Experimental ACP composite efficiently established mineral ion transfer throughout the body of the lesions and restored the mineral lost due to acid attack. It can be considered a useful adjuvant for the control of caries in orthodontic applications.
Sigma-Aldrich
Bisphenol A ethoxylate dimethacrylate, average Mn ~1,700, EO/phenol 15, contains 200 ppm MEHQ as inhibitor