MilliporeSigma
  • Characterization of D-tagatose-3-epimerase from Rhodobacter sphaeroides that converts D-fructose into D-psicose.
Characterization of D-tagatose-3-epimerase from Rhodobacter sphaeroides that converts D-fructose into D-psicose.
Biotechnology letters (2009-02-12)
Longtao Zhang, Wanmeng Mu, Bo Jiang, Tao Zhang
ABSTRACT
A non-characterized gene, previously proposed as the D-tagatose-3-epimerase gene from Rhodobacter sphaeroides, was cloned and expressed in Escherichia coli. Its molecular mass was estimated to be 64 kDa with two identical subunits. The enzyme specificity was highest with D-fructose and decreased for other substrates in the order: D-tagatose, D-psicose, D-ribulose, D-xylulose and D-sorbose. Its activity was maximal at pH 9 and 40 degrees C while being enhanced by Mn(2+). At pH 9 and 40 degrees C, 118 g D-psicose l(-1) was produced from 700 g D-fructose l(-1) after 3 h.
Sigma-Aldrich
D-Ribulose solution, ~1 M in H2O, ≥97.0% (HPLC)
Sigma-Aldrich
D-Psicose, ≥95% (HPLC)