MilliporeSigma
  • Efficient solid-phase synthesis of sulfotyrosine peptides using a sulfate protecting-group strategy.
Efficient solid-phase synthesis of sulfotyrosine peptides using a sulfate protecting-group strategy.
Angewandte Chemie (International ed. in English) (2009-02-05)
Ahmed M Ali, Scott D Taylor
ABSTRACT
Double protection: Efficient Fmoc-based solid-phase synthesis (SPPS) of sulfotyrosine (sY) peptides is achieved by incorporating the sY residue(s) as a dichlorovinyl-protected (DCV) sulfodiester(s) and using 2-methylpiperidine for Fmoc removal. After removal of the other protecting groups, the DCV group could be cleaved by mild hydrogenolysis giving the sY peptides in good yield.
Sigma-Aldrich
2-Methylpiperidine, 98%