MilliporeSigma
  • Maintenance of an acidic stratum corneum prevents emergence of murine atopic dermatitis.
Maintenance of an acidic stratum corneum prevents emergence of murine atopic dermatitis.
The Journal of investigative dermatology (2009-01-30)
Yutaka Hatano, Mao-Qiang Man, Yoshikazu Uchida, Debra Crumrine, Tiffany C Scharschmidt, Esther G Kim, Theodora M Mauro, Kenneth R Feingold, Peter M Elias, Walter M Holleran
ABSTRACT
Neutralization of stratum corneum (SC) adversely impacts key epidermal functions, including permeability barrier homeostasis and SC integrity. Conversely, acidification of SC improves these functions in developmentally impaired (neonatal or aged) skin, and enhances function in normal skin. Hence, we hypothesized that acidification could alter the course of inflammatory dermatoses, which invariably exhibit an increased SC pH. Maintenance of a low pH by topical applications of the polyhydroxyl acid, lactobionic acid, during the repeated-challenge phase inhibited the development of oxazolone-induced atopic dermatitis (AD). Neither gross/histological dermatitis nor altered barrier function developed, and emergence of epidermal hyperplasia was prevented; however, cytokine generation decreased. Acidification also largely normalized the development of hapten-induced changes in eosinophil/mast cell densities, density of chemoattractant receptor-homologous molecule expressed on TH2-positive lymphocytes, and serum IgE levels. The pH-induced improvement in barrier function most likely accounts for the anti-inflammatory activity, which could be further attributed to normalization of both lamellar body secretion and lamellar bilayer formation. Acidification of SC alone substantially prevents development of barrier abnormalities and downstream immune abnormalities during the elicitation phase of murine AD. These results provide direct evidence for the "outside-inside" pathogenesis of AD and further suggest that maintenance of an acidic SC pH could prevent the emergence of AD in humans.
Sigma-Aldrich
Lactobionic acid, 97% (TLC)
Sigma-Aldrich
Lactobionic acid, ≥97% (TLC), cell impermeant agent
Lactobionic acid, European Pharmacopoeia (EP) Reference Standard