MilliporeSigma
  • Catalytic activity-independent pathway is involved in phospholipase A(2)-induced apoptotic death of human leukemia U937 cells via Ca(2+)-mediated p38 MAPK activation and mitochondrial depolarization.
Catalytic activity-independent pathway is involved in phospholipase A(2)-induced apoptotic death of human leukemia U937 cells via Ca(2+)-mediated p38 MAPK activation and mitochondrial depolarization.
Toxicology letters (2009-01-03)
Wen-Hsin Liu, Pei-Hsiu Kao, Yi-Ling Chiou, Shinne-Ren Lin, Ming-Jung Wu, Long-Sen Chang
ABSTRACT
In view of the controversial role of catalytic activity on the cytotoxicity of phospholipase A(2) (PLA(2)), the present study is conducted to explore whether PLA(2) induces apoptotic process of human leukemia U937 cells through catalytic activity-independent pathway. Modification of His-48 (according to the sequence alignment with porcine pancreatic PLA(2)) with p-bromophenacyl bromide (BPB) caused over 99.9% drop in enzymatic activity Naja naja atra PLA(2). It was found that BPB-PLA(2)-induced apoptotic death of U937 cells was associated with mitochondrial depolarization, modulation of Bcl-2 family members, cytochrome c release and activation of caspases 9 and 3. Upon exposure to BPB-PLA(2), elevation of intracellular Ca(2+) levels and p38 MAPK activation were observed in U937 cells. Pretreatment with BAPTA-AM (Ca(2+) chelator) and nifedipine (L-type Ca(2+) channel blocker) abrogated Ca(2+) increase and p38 MAPK activation, and rescued viability of BPB-PLA(2)-treated U937 cells. BPB-PLA(2)-induced dissipation of mitochondrial membrane potential and down-regulation of Bcl-2 were suppressed by SB202190 (p38MAPK inhibitor). Although PLA(2) mutants in which His-48 and Asp-49 were substituted by Ala and Lys, respectively, did not display detectable PLA(2) activity, they induced death of U937 cells. The signaling pathway of PLA(2) mutants in inducing cell death was indistinguishable from that of BPB-PLA(2). Taken together, our data indicate that catalytic activity-independent pathway is involved in PLA(2)-induced apoptotic death of human leukemia U937 cells via mitochondria-mediated death pathway triggering by Ca(2+)-mediated p38 MAPK activation.
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%
Supelco
2,4′-Dibromoacetophenone, LiChropur, ≥99.0% (HPLC)