MilliporeSigma
  • Targetry of SrCO(3) on a copper substrate by sedimentation method for the cyclotron production no-carrier-added (86)Y.
Targetry of SrCO(3) on a copper substrate by sedimentation method for the cyclotron production no-carrier-added (86)Y.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2008-12-27)
M Sadeghi, A Zali, M Aboudzadeh, P Sarabadani, G Aslani, A Majdabadi
ABSTRACT
Strontium carbonate deposition on copper substrate was carried out by the sedimentation method in order to produce yttrium-86. Natural strontium carbonate thick layer was prepared with 480 mg SrCO(3), 220 mg ethyl cellulose, and 7.5 mL acetone. This optimum condition is a result of several repeated experiments with different amount of ethyl cellulose and acetone. Target quality control was done by SEM photomicrograph and thermal shock test. The deposited target was irradiated at 30 microA current and 15 MeV proton beam for 12 min.
Sigma-Aldrich
Strontium carbonate, ≥98%
Sigma-Aldrich
Strontium carbonate, ≥99.9% trace metals basis
Sigma-Aldrich
Strontium carbonate, 99.995% trace metals basis