MilliporeSigma
  • Analysis of capsaicin and dihydrocapsaicin in peppers and pepper sauces by solid phase microextraction-gas chromatography-mass spectrometry.
Analysis of capsaicin and dihydrocapsaicin in peppers and pepper sauces by solid phase microextraction-gas chromatography-mass spectrometry.
Journal of chromatography. A (2008-12-23)
Araceli Peña-Alvarez, Erika Ramírez-Maya, Luís Angel Alvarado-Suárez
ABSTRACT
A simple method for the analysis of capsaicin and dihydrocapsaicin in peppers and pepper sauces by solid phase microextraction-gas chromatography-mass spectrometry has been developed. A novel device was designed for direct extraction solid phase microextraction in order to avoid damage to the fiber. The analysis was performed without derivatization for the gas chromatography-mass spectrometry analysis. Selection fiber, extraction temperature, extraction time and pH, were optimized. The method was linear in the range 0.109-1.323 microg/mL for capsaicin and 0.107-1.713 microg/mL for dihydrocapsaicin with correlation coefficient up to r=0.9970 for both capsaicinoids. The precision of the method was less than 10%. The method was applied to the analysis of 11 varieties of peppers and four pepper sauces. A broad range of capsaicin (55.0-25 459 microg/g) and dihydrocapsaicin (93-1 130 microg/g) was found in the pepper and pepper sauces samples (4.3-717.3 and 1.0-134.8 microg/g), respectively.
Sigma-Aldrich
Dihydrocapsaicin, from Capsicum sp., ≥85%
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler