MilliporeSigma
  • Rapid three-step one-pot microwave-assisted synthesis of 2,5-dioxo-1,2,5,6,7,8-hexahydro-3-quinolinecarbonitrile library.
Rapid three-step one-pot microwave-assisted synthesis of 2,5-dioxo-1,2,5,6,7,8-hexahydro-3-quinolinecarbonitrile library.
Journal of combinatorial chemistry (2008-12-09)
Sergey A Yermolayev, Nikolay Yu Gorobets, Sergey M Desenko
ABSTRACT
Many well-known drugs contain 2-pyridone and 2-quinolone scaffolds. In the current paper, a one-pot three-step microwave-assisted method for the synthesis of N1-substituted 2,5-dioxo-1,2,5,6,7,8-hexahydro-3-quinolinecarbonitrile derivatives was developed. Employing this protocol, we quickly generated 105 compounds library from 1,3-cyclohexanediones, dimethylformamide dimethylacetal, and various cyanacetamides.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%