MilliporeSigma
  • Comparative bioavailability of two estazolam tablet formulations in Indonesian healthy volunteers.
Comparative bioavailability of two estazolam tablet formulations in Indonesian healthy volunteers.
Arzneimittel-Forschung (2008-11-26)
Yahdiana Harahap, Lucy Sasongko, Budi Prasaja, Ega Indriati, Windy Lusthom, Lipin
ABSTRACT
To compare the bioavailability of two estazolam (CAS 29975-16-4) tablet formulations (Estalin 2 mg tablets as test formulation and 2 mg tablets of the originator product as reference formulation). The study was conducted according to an open label, randomized two-way cross-over design with a two-week washout period. Twenty-four subjects received each of the two estazolam formulations. Blood samples for pharmacokinetic profiling were taken up to 72 h after drug administration in fasting condition. Plasma concentrations of estazolam were determined with a validated HPLC method with ultraviolet detection. Pharmacokinetic parameters were calculated from observed plasma concentration-time profiles. The mean AUC(0-t), AUC(0-infinity) and Cmax were 2581.38 ng x h/mL, 2934.37 ng x h/mL and 95.25 ng/mL, respectively for the test formulation and 2835.75 ng x h/ mL, 3207.73 ng x h/mL and 99.32 ng/mL, respectively, for the reference formulation. The median Tmax for both formulations was 1 h. The point estimates and 90% confidence Intervals for AUC(0-t), AUC(0-infinity) and Cmax were 91.03% (87.48-94.72%), 91.48% (86.67-96.55%) and 95.90% (92.60-99.31%) respectively, satisfying the bloequivalence criteria of the European Committee for Proprietary Medicinal Products and the US Food and Drug Administration guidelines. These results indicate that two formulations of estazolam are bioequivalent and, thus, may be prescribed interchangeably.
Product Description
Supelco
Estazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®