MilliporeSigma
  • Synthesis of regioselectively sulfated xylodextrins and crystal structure of sodium methyl beta-D-xylopyranoside 4-O-sulfate hemihydrate.
Synthesis of regioselectively sulfated xylodextrins and crystal structure of sodium methyl beta-D-xylopyranoside 4-O-sulfate hemihydrate.
Carbohydrate research (2008-11-11)
Beatriz Abad-Romero, Kurt Mereiter, Herbert Sixta, Andreas Hofinger, Paul Kosma
ABSTRACT
Methyl xylobioside and methyl xylotrioside were prepared from the peracetylated anomeric xylosyl trichloroacetimidates by reaction with methanol followed by Zemplén deacetylation. Methyl beta-D-xylopyranoside, methyl beta-D-xylobioside and methyl beta-D-xylotrioside were subjected to treatment with dibutyltin oxide followed by reaction with the trimethylamine/sulfur trioxide complex in tetrahydrofuran. This way, preferential sulfation of the terminal 4-hydroxy group at the nonreducing xylopyranosyl unit was achieved. In addition, partial sulfation at position 2 of the distal xylose unit was observed. The substitution pattern was derived from NMR spectroscopic data and was confirmed by the X-ray structure determination of sodium methyl beta-D-xylopyranoside 4-O-sulfate. The compound crystallized as a hemihydrate in a triclinic lattice of space group P1 and possesses a pseudomonoclinic 2D supramolecular structure. The sulfation of free pentose oligomers via their intermediate stannylene acetals may thus be exploited to generate biologically active oligosaccharides for biomedical applications.
Sigma-Aldrich
Methyl sulfate sodium salt