MilliporeSigma
Electron transfer through a stable phenanthrenyl pair in DNA.
Chemical communications (Cambridge, England) (2008-11-06)
Nikolay A Grigorenko, Christian J Leumann
ABSTRACT
Within an oligonucleotide duplex, excess electron transfer from a excited 5-(pyren-1-yl)uridine through an internal stacked phenanthrenyl pair to 5-bromouridine as an electron acceptor was observed.
Sigma-Aldrich
5-Bromouridine, 98%