MilliporeSigma
  • Selenocystine induces S-phase arrest and apoptosis in human breast adenocarcinoma MCF-7 cells by modulating ERK and Akt phosphorylation.
Selenocystine induces S-phase arrest and apoptosis in human breast adenocarcinoma MCF-7 cells by modulating ERK and Akt phosphorylation.
Journal of agricultural and food chemistry (2008-10-31)
Tianfeng Chen, Yum-Shing Wong
ABSTRACT
Selenocystine (SeC) is a nutritionally available selenoamino acid with selective anticancer effects on a number of human cancer cell lines. The present study shows that SeC inhibited the proliferation of human breast adenocarcinoma MCF-7 cells in a time- and dose-dependent manner, through the induction of cell cycle arrest and apoptotic cell death. SeC-induced S-phase arrest was associated with a marked decrease in the protein expression of cyclins A, D1, and D3 and cyclin-dependent kinases (CDKs) 4 and 6, with concomitant induction of p21waf1/Cip1, p27Kip1, and p53. Exposure of MCF-7 cells to SeC resulted in apoptosis as evidenced by caspase activation, PARP cleavage, and DNA fragmentation. SeC treatment also triggered the activation of JNK, p38 MAPK, ERK, and Akt. Inhibitors of ERK (U0126) and Akt (LY294002), but not JNK (SP600125) and p38 MAPK (SB203580), suppressed SeC-induced S-phase arrest and apoptosis in MCF-7 cells. The findings establish a mechanistic link between the PI3K/Akt pathway, MAPK pathway, and SeC-induced cell cycle arrest and apoptosis in MCF-7 cells.
Sigma-Aldrich
Seleno-L-cystine, 95%
Sigma-Aldrich
Seleno-DL-cystine, powder