MilliporeSigma
  • Biochemical properties of group G streptococci isolated from cats and man.
Biochemical properties of group G streptococci isolated from cats and man.
Journal of medical microbiology (1991-09-01)
J C Reitmeyer, R K Guthrie, J H Steele
ABSTRACT
The biochemical characteristics of group G streptococci isolated from cats were markedly similar to the characteristics of group G streptococci from man. Both cat and human isolates of group G streptococci were also very similar in biochemical characteristics to group A streptococci so that to identify the source of group G streptococci by biochemical reactions is not a reliable procedure. The group G streptococci found in many cats could be pathogenic to man, since their physiological and biological characteristics are very similar to those of group A streptococci.
Sigma-Aldrich
Indoxyl acetate, ≥95% (GC)