MilliporeSigma
  • Complexes of 2-acetyl-gamma-butyrolactone and 2-furancarbaldehyde thiosemicarbazones: antibacterial and antifungal activity.
Complexes of 2-acetyl-gamma-butyrolactone and 2-furancarbaldehyde thiosemicarbazones: antibacterial and antifungal activity.
Journal of inorganic biochemistry (2008-10-25)
María C Rodríguez-Argüelles, Patricia Tourón-Touceda, Roberto Cao, Ana M García-Deibe, Paolo Pelagatti, Corrado Pelizzi, Franca Zani
ABSTRACT
Cobalt, nickel, copper and zinc coordination compounds of two thiosemicarbazones with general composition ML(2) (L: monodeprotonated ligand corresponding to 2-acetyl-gamma-butyrolactone thiosemicarbazone, HL(1), and 2-furancarbaldehyde thiosemicarbazone, HL(2)) and also complexes with general composition MCl(2)(HL(2)) were synthesized (except [NiCl(2)(HL(2))] and [Co(L(2))(2)]). The interaction of CuCl(2) with HL(2) gave [CuCl(HL(2))], a copper(I) complex. The ligands and metal complexes were characterized by IR, (1)H and (13)C NMR spectroscopy, and magnetic susceptibility measurements. The crystal structure of [Ni(L(2))(2)]x 2dmso was determined and a trans-square planar coordination of the two kappa(2)-N,S chelate rings forming polymeric strips through H-bonds with dmso was observed. Actually, in all the reported complexes both ligands behaved as kappa(2)-N,S chelates, except in the case of [Co(L(1))(2)] in which HL(1) is tridentate kappa(3)-N,S,O. The antimicrobial properties of all compounds were studied using a wide spectrum of bacterial and fungal strains. The copper complexes of HL(2) were the most active against all strains, including dermatophytes and phytopathogenic fungi. Most of the studied compounds, especially [Cu(L(1))(2)], presented good activity against Haemophilus influenzae, a very harmful bacterium to humans.
Sigma-Aldrich
2-Furoic acid, 98%