MilliporeSigma
  • Study of the reaction between methyl 4-nitrobenzenesulfonate and bromide ions in mixed single-chain-gemini micellar solutions: kinetic evidence for morphological transitions.
Study of the reaction between methyl 4-nitrobenzenesulfonate and bromide ions in mixed single-chain-gemini micellar solutions: kinetic evidence for morphological transitions.
Journal of colloid and interface science (2008-10-09)
María del Mar Graciani, Amalia Rodríguez, María Luisa Moyá
ABSTRACT
The reaction between methyl 4-nitrobenzenesulfonate and bromide ions has been studied in mixed single-chain-gemini micellar solutions of n-dodecyltrimethylammonium bromide, DTAB, and dodecyl tricosaoxyethylene glycol ether, Brij(35), with alkanediyl-alpha-omega-bis(dodecyldimethylammonium) bromide, 12-s-12,2Br(-) (s=3,4,5). Kinetic micellar effects show that an increase in the solution mole fraction of the single-chain surfactant, X(single-chain), results in a diminution of the mixed micelles tendency to form spherocylindrical aggregates upon increasing surfactant concentration. The dependence of the surfactant concentration at which the sphere-to-rod transition occurs, C(*), on X(single-chain) showed through kinetic data was in agreement with results obtained by means of fluorescence measurements.
Sigma-Aldrich
Methyl 4-nitrobenzenesulfonate, 99%