MilliporeSigma
Novel chalcones derived from 2-chloro-3-formyl-6-methylquinoline.
Acta crystallographica. Section C, Crystal structure communications (2008-10-08)
Umar Farooq Rizvi, Hamid Latif Siddiqui, Saeed Ahmad, Matloob Ahmad, Masood Parvez
ABSTRACT
Molecules of (E)-3-(2-chloro-6-methylquinolin-3-yl)-1-(5-iodo-2-thienyl)prop-2-en-1-one, C(17)H(11)ClINOS, (I), and (E)-3-(2-chloro-6-methylquinolin-3-yl)-1-(5-methyl-2-furyl)prop-2-en-1-one, C(18)H(14)ClNO(2), (II), adopt conformations slightly twisted from coplanarity. Both structures are devoid of classical hydrogen bonds. However, nonclassical C-H...O/N interactions [with C...O = 3.146 (5) A and C...N = 3.487 (3) A] link the molecules into chains extended along the b axis in (I) and form dimers with an R(2)(2)(8) motif in (II). The structural analysis of these compounds provides an insight into the correlation between molecular structures and intermolecular interactions in compounds for drug development.
Sigma-Aldrich
6-Methylquinoline, ≥98%, FG
Sigma-Aldrich
6-Methylquinoline, 98%