MilliporeSigma
  • The putrescine analogue 1,4-diamino-2-butanone affects polyamine synthesis, transport, ultrastructure and intracellular survival in Leishmania amazonensis.
The putrescine analogue 1,4-diamino-2-butanone affects polyamine synthesis, transport, ultrastructure and intracellular survival in Leishmania amazonensis.
Microbiology (Reading, England) (2008-10-04)
Marcos A Vannier-Santos, Diego Menezes, Marcus F Oliveira, Fernando G de Mello
ABSTRACT
Polyamines are important regulators of growth and differentiation in a variety of cells, including parasitic protozoa. Promastigotes of Leishmania species have high levels of putrescine and spermidine and their growth can be inhibited by polyamine biosynthesis antagonists. The putrescine analogue 1,4-diamino-2-butanone (DAB) is microbicidal against Tritrichomonas foetus and Trypanosoma cruzi, so we tested its effects on Leishmania amazonensis proliferation, viability, organization, putrescine transport and synthesis as well as in vitro infectivity. DAB impaired promastigote proliferation dose-dependently (IC(50) 144 microM) and the parasite putrescine concentration was reduced by nearly 50 %. This analogue markedly inhibited both ornithine decarboxylase activity and [H(3)]putrescine uptake by promastigotes. Pre-treatment with DAB for 24 h led to compensatory enhancement of putrescine uptake, indicating an adaptive mechanism in DAB-treated parasites. Remarkably, DAB caused mitochondrial damage, assessed by transmission electron microscopy, and 3 h treatment with 1 mM DAB enhanced lipid peroxidation, whereas incubation with 10 mM DAB or for 24 h resulted in decreased peroxidation levels in the parasites. This effect was probably due to the loss of mitochondrial function, demonstrated by the diminished reduction of 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), not observed in macrophages. Murine macrophages infected with L. amazonensis amastigotes treated with DAB had parasite loads significantly (P<0.05) lower than controls, presumably due to interference with putrescine uptake and/or synthesis. These results suggest that putrescine may be involved in leishmanial survival, possibly by maintaining the parasite's mitochondrial function. The use of analogues to interfere with polyamine/diamine synthesis and transport may shed light on its function in intracellular parasite survival and lead to identification of new targets for leishmaniasis chemotherapy.