MilliporeSigma
  • Semisynthesis and antitumoral activity of 2-acetylfuranonaphthoquinone and other naphthoquinone derivatives from lapachol.
Semisynthesis and antitumoral activity of 2-acetylfuranonaphthoquinone and other naphthoquinone derivatives from lapachol.
Bioorganic & medicinal chemistry letters (2008-10-03)
Kenneth O Eyong, Ponminor S Kumar, Victor Kuete, Gabriel N Folefoc, Ephriam A Nkengfack, Sundarababu Baskaran
ABSTRACT
Ozonolysis of lapachol (1), resulting in an unusual formation of a potent antitumor agent 2-acetylfuranonaphthoquinone (3) along with the expected aldehyde 6, is described. The reaction of lapachol (1) with CAN in dry acetonitrile leading to biologically active furanonaphthoquinones is also reported. The antitumoral activity of the tested compounds on human DU-145 prostate carcinoma cells was evaluated following XTT assay. The results revealed that 2-(1-methylethenyl)-2,3-dihydronaphtho[2,3-b]furan-4,9-dione (5), beta-lapachone (10) and dehydro-beta-lapachone diacetate (11) showed 100% inhibition at 25 microg/ml. All the tested samples showed dose-dependent activity.
Sigma-Aldrich
Lapachol, 98%