MilliporeSigma
  • Electrophysiological features of late-onset transthyretin Met30 familial amyloid polyneuropathy unrelated to endemic foci.
Electrophysiological features of late-onset transthyretin Met30 familial amyloid polyneuropathy unrelated to endemic foci.
Journal of neurology (2008-09-30)
Haruki Koike, Yuichi Kawagashira, Masahiro Iijima, Masahiko Yamamoto, Naoki Hattori, Fumiaki Tanaka, Masaaki Hirayama, Yukio Ando, Shu-ichi Ikeda, Gen Sobue
ABSTRACT
Through the development of gene diagnostic techniques, late-onset transthyretin Met30-associated familial amyloid polyneuropathy (FAP TTR Met30) has been shown to be more prevalent than is generally believed. To examine the electrophysiological features of late-onset FAP TTR Met30 unrelated to endemic foci. Nerve conduction findings in 44 cases with an onset of more than 50 years of age in a non-endemic area were assessed and compared with findings from 21 earlier-onset cases related to endemic foci. The extent of the reduction of the compound muscle action potential and, especially, the sensory nerve action potential was more profound in the late-onset group even when the decline of these indices with aging in normal control subjects was taken into account. The feature of predominant lower-limb involvement seemed to be more conspicuous in the late-onset group. Electrophysiological indices tended to be aggravated as the duration of neuropathic symptoms increased in the early-onset group, while most of these indices in the lateonset group did not show this correlation. A slowing of conduction velocity and a prolongation of distal latency, which suggests demyelination, were conspicuous in some patients. Pathologically, a predominant loss of small-fibers was not conspicuous in sural nerve biopsy specimens from late-onset patients. Large myelinated fiber density showed a negative correlation with the disease duration in early-onset cases, but not in late-onset cases. Electrophysiological differences between late- and early-onset cases were present, probably reflecting the different underlying pathogenic mechanisms of neuropathy. The demyelinating feature does not exclude the possibility of this disease.
Sigma-Aldrich
Prealbumin from human plasma, lyophilized powder