MilliporeSigma
  • Thermochemistry of 2- and 3-thiopheneacetic acids: calorimetric and computational study.
Thermochemistry of 2- and 3-thiopheneacetic acids: calorimetric and computational study.
The journal of physical chemistry. A (2008-09-26)
Manuel Temprado, María Victoria Roux, Pilar Jiménez, Concepción Foces-Foces, Rafael Notario
ABSTRACT
The enthalpies of formation in the condensed and gas states, Delta f H m degrees (cd) and Delta f H m degrees (g), of 2- and 3-thiopheneacetic acids were derived from their respective enthalpies of combustion in oxygen, measured by a rotating bomb calorimeter, and the variation of vapor pressure with temperature determined by the Knudsen effusion technique. Theoretical calculations at the G3 level were performed, and a study on molecular and electronic structure of the compounds has been carried out. Calculated Delta f H m degrees (g) values using atomization and isodesmic reactions are compared with the experimental data. Experimental and theoretical results show that the 3-thiopheneacetic acid is thermodynamically more stable than the 2-isomer.
Sigma-Aldrich
2-Thiopheneacetic acid, 98%