MilliporeSigma
Hydrous zinc halide-catalyzed aminosulfonation of hydrocarbons.
Chemical communications (Cambridge, England) (2008-09-20)
Biswajit Kalita, Angus A Lamar, Kenneth M Nicholas
ABSTRACT
Benzylic and allylic hydrocarbons are selectively converted to the corresponding sulfonamides by a ZnBr2-H2O-catalyzed reaction with PhI=NTs; saturated adamantane is aminosulfonated at the tertiary C-H bond.
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis