MilliporeSigma
  • Graft polymerization of wood sawdust and peat with ethylene carbonate. A novel method for the preparation of supports with enhanced mechanical properties to be used in biofiltration of organic vapors.
Graft polymerization of wood sawdust and peat with ethylene carbonate. A novel method for the preparation of supports with enhanced mechanical properties to be used in biofiltration of organic vapors.
Bioresource technology (2008-09-02)
O Hernández-Meléndez, J Peydecastaing, E Bárzana, C Vaca-Garcia, M Hernández-Luna, M E Borredon
ABSTRACT
The graft polymerization reaction between ethylene carbonate (EC) and scots pine sawdust (SPS) or peat moss (PM) offers a solvent-free approach to the simple and inexpensive aliphatic derivatization of these lignocellulosic fibers. This reaction was studied with liquid or vapor EC phases in three different reactor configurations: batch stirred (BSR), semi-continuous stirred (SSR) and continuous tubular in the gas phase (CVTR). The use of a vapor phase allowed a satisfactory grafting yield and minimal production of non-grafted polyol by-products. The crosslinking agent 4,4'-methylenebis(phenylisocyanate) (MDI) achieved superior characteristics to form shaped tablets resistant to water disaggregation, a high water retention capacity and high compression strength, characteristics that conventional organic supports like PM or PM-polyurethane foam mixtures used in biofiltration of waste gases do not completely possess.
Sigma-Aldrich
Ethylene carbonate, 98%