MilliporeSigma
  • Steroids from the roots of Asparagus officinalis and their cytotoxic activity.
Steroids from the roots of Asparagus officinalis and their cytotoxic activity.
Journal of integrative plant biology (2008-08-21)
Xue-Feng Huang, Yu-Ying Lin, Ling-Yi Kong
ABSTRACT
One new (Sarsasapogenin O) and seven known steroids were isolated from the roots of Asparagus officinalis L. Their structures were elucidated on the basis of spectroscopic analysis, including various 2D-NMR techniques, hydrolysis, and by comparison of spectral data of known compounds. These compounds together with nine steroids which were previously isolated from this plant, were tested for cytotoxic activity. Among them, eight compounds displayed significant cytotoxicities against human A2780, HO-8910, Eca-109, MGC-803, CNE, LTEP-a-2, KB and mouse L1210 tumor cells.
Sigma-Aldrich
Sarsasapogenin, ≥98%