MilliporeSigma
  • Antifungal activity of the termite alkaloid norharmane against the mycelial growth of Metarhizium anisopliae and Aspergillus nomius.
Antifungal activity of the termite alkaloid norharmane against the mycelial growth of Metarhizium anisopliae and Aspergillus nomius.
Journal of invertebrate pathology (2008-08-16)
Thomas Chouvenc, Nan-Yao Su, Monica I Elliott
ABSTRACT
Antifungal activity of norharmane, a beta-carboline alkaloid found in termites (Isoptera, Rhinotermitidae) was tested against two entomopathogenic fungi, Metarhizium anisopliae and Aspergillus nomius. It was determined that, at physiological concentration (10 microg ml(-1)), norharmane had no significant effect on A. nomius mycelial growth rate but reduced M. anisopliae growth rate by 11.9%. Contrary to previous findings, we suggest that norharmane has a limited role in disease resistance against fungal pathogens in individual subterranean termites, and we discuss the potential role of this chemical at a colony level.
Sigma-Aldrich
Norharmane, crystalline