MilliporeSigma
  • Reactions of copper(II)-H2O2 adducts supported by tridentate bis(2-pyridylmethyl)amine ligands: sensitivity to solvent and variations in ligand substitution.
Reactions of copper(II)-H2O2 adducts supported by tridentate bis(2-pyridylmethyl)amine ligands: sensitivity to solvent and variations in ligand substitution.
Inorganic chemistry (2008-08-14)
Atsushi Kunishita, Joseph D Scanlon, Hirohito Ishimaru, Kaoru Honda, Takashi Ogura, Masatatsu Suzuki, Christopher J Cramer, Shinobu Itoh
ABSTRACT
The copper(II) complexes 1(H) and 1(Ar(X)), supported by the N,N-di(2-pyridylmethyl)benzylamine tridentate ligand (L(H)) or its derivatives having m-substituted phenyl group at the 6-position of pyridine donor groups (L(Ar(X))), have been prepared, and their reactivity toward H2O2 has been examined in detail at low temperature. Both copper(II) complexes exhibited a novel reactivity in acetone, giving 2-hydroxy-2-hydroperoxypropane (HHPP) adducts 2(H) and 2(Ar(X)), respectively. From 2(Ar(X)), an efficient aromatic ligand hydroxylation took place to give phenolate-copper(II) complexes 4(Ar(X)). Detailed spectroscopic and kinetic analyses have revealed that the reaction proceeds via an electrophilic aromatic substitution mechanism involving copper(II)-carbocation intermediates 3(Ar(X)). Theoretical studies at the density functional theory (DFT) level have strongly implicated conjugate acid/base catalysis in the O-O bond cleavage and C-O bond formation steps that take the peroxo intermediate 2(Ar(X)) to the carbocation intermediate 3(Ar(X)). In contrast to the 2(Ar(X)) cases, the HHPP-adduct 2(H) reacted to give a copper(II)-acetate complex [Cu(II)(L(H))(OAc)](ClO4) (6(H)), in which one of the oxygen atoms of the acetate co-ligand originated from H2O2. In this case, a mechanism involving a Baeyer-Villiger type 1,2-methyl shift from the HHPP-adduct and subsequent ester hydrolysis has been proposed on the basis of DFT calculations; conjugate acid/base catalysis is implicated in the 1,2-methyl shift process as well. In propionitrile, both 1(H) and 1(Ar(X)) afforded simple copper(II)-hydroperoxo complexes LCu(II)-OOH in the reaction with H2O2, demonstrating the significant solvent effect on the reaction between copper(II) complexes and H2O2.
Product Description
Supelco
Propionitrile, analytical standard
Sigma-Aldrich
Propionitrile, purum, ≥99.0% (GC)
Sigma-Aldrich
Propionitrile, 99%