MilliporeSigma
Synthetic studies on formamidopyrimidines related to clofarabine.
Nucleosides, nucleotides & nucleic acids (2008-08-13)
William R Cantrell, Dennis Lovett, Tracy Engles, Bruce Anderson, William E Bauta, Paul C Wolstenholme-Hogg
ABSTRACT
Degradation of clofarabine (3) in 0.9% saline solution at 100 degrees C afforded three degradation products which were determined to be formamidopyrimidines 4-6. Compounds 4 and 5 were assigned as C(1') anomers on the basis of one-dimensional and two-dimensional NMR experiments, whereas 6 was found to be the formamidopyrimidine lacking the sugar moiety. An improved procedure for the synthesis of formamidopyrimidines was developed, wherein benzoylated clofarabine (11) was treated with allyl chloroformate, followed by deprotection of the alloc group with catalytic Pd(PPh(3))(4) and dimedone. A synthesis of compound 6 from 4 is also described.
Sigma-Aldrich
Allyl chloroformate, 97%