MilliporeSigma
  • Degradation and metabolite formation of 17beta-estradiol-3-sulphate in New Zealand pasture soils.
Degradation and metabolite formation of 17beta-estradiol-3-sulphate in New Zealand pasture soils.
Environment international (2008-08-13)
Frank F Scherr, Ajit K Sarmah, Hong J Di, Keith C Cameron
ABSTRACT
Estrogens-sulphates such as 17beta-estradiol-3-sulphate and estrone-3-sulphate are excreted by livestock in the urine. These conjugates are precursors to the free counterparts 17beta-estradiol and estrone, which are endocrine disrupting chemicals. In this study microcosm laboratory experiments were conducted in three pasture soils from New Zealand to study the aerobic degradation and metabolite formation kinetics of 17beta-estradiol-3-sulphate at three different incubation temperatures. The degradation of 17beta-estradiol-3-sulphate followed a first-order kinetic and the temperature dependence of the rate constants was sufficiently described by the Arrhenius equation. Degradation was different between the three investigated soils and the rate constants across the soils were significantly correlated to the arylsulphatase activity at 7.5 and 15 degrees C. Estrone-3-sulphate and 17beta-estradiol were identified as primary metabolites and estrone as a secondary metabolite. Results suggest arylsulphatase activity originating from soil microbial biomass is the main driver for the degradation of 17beta-estradiol-3-sulphate.
Sigma-Aldrich
β-Estradiol 3-sulfate sodium salt, ≥93%