MilliporeSigma
  • Pharmacokinetics of lidocaine and its metabolite in peridural anesthesia administered to pregnant women with gestational diabetes mellitus.
Pharmacokinetics of lidocaine and its metabolite in peridural anesthesia administered to pregnant women with gestational diabetes mellitus.
European journal of clinical pharmacology (2008-08-06)
Elaine Christine Dantas Moisés, Luciana de Barros Duarte, Ricardo de Carvalho Cavalli, Maria Paula Marques, Vera Lúcia Lanchote, Geraldo Duarte, Sérgio Pereira da Cunha
ABSTRACT
Peridural blockade with lidocaine, bupivacaine, and fentanyl is an anesthetic procedure extensively used in obstetrics, justifying the pharmacokinetic study of these drugs during labor. To investigate the influence of the physiopathological changes of gestational diabetes mellitus (GDM) on the pharmacokinetics of lidocaine and its metabolite monoethylglycinexylidide (MEGX) in pregnant women subjected to peridural anesthesia. Ten normal pregnant women (group 1) and six pregnant women with GDM (group 2) were studied, all of them at term. The patients received 200 mg 2% lidocaine hydrochloride without a vasoconstrictor by the peridural locoregional route. Maternal blood samples were collected at predetermined times for the analysis of lidocaine and MEGX by chromatography and pharmacokinetic analysis. The median pharmacokinetic parameters of lidocaine for groups 1 and 2 (P </= 0.05), respectively, were as follows: for Cmax 879.11 and 1,145.58 ng/ml, AUC(0-infinity) 256.01 and 455.95 mug min(-1) ml(-1), Cl/f/kg 10.61 and 5.64 ml min(-1) kg(-1), and Vd/f/kg 3.26 and 2.19 L/kg. The median pharmacokinetic parameters of MEGX for groups 1 and 2 (P </= 0.05), respectively, were as follows: for Cmax 82.71 and 141.38 ng/ml, Tmax 44.71 and 193.14 min, t(1/2)alpha 7.64 and 59.77 min, alpha 0.097 and 0.012/min, and AUC(0-infinity) 29.91 and 108.23 mug min(-1) ml(-1). The present data permit us to conclude that the apparent clearance of lidocaine and MEGX was reduced in diabetic patients compared to normal women, suggesting that GDM inhibits the CYP1A2/CYP3A4 isoforms responsible for the metabolism of this drug and its metabolite.
Sigma-Aldrich
MEGX, ≥95% (HPLC)