MilliporeSigma
  • Determination of reactions between free radicals and selected Chilean wines and transition metals by ESR and UV-vis technique.
Determination of reactions between free radicals and selected Chilean wines and transition metals by ESR and UV-vis technique.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-08-05)
Mónica Espinoza, Claudio Olea-Azar, Hernán Speisky, Jorge Rodríguez
ABSTRACT
Four different types of Chilean wines (Cabernet Sauvignon, Merlot, Carmenere and Syrah) were selected and examined in their free radical scavenging capacities by electron spin resonance (ESR) and spectrophotometric methods. The free radical scavenging properties were evaluated against 2,2-diphenyl-1-picrylhydrazyl (DPPH*) radical, 2,6-di-tert-butyl-alpha-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy (Galvinoxyl) radical and hydroxyl radical (HO*). The possible effect on these scavenging properties of added transition metals to these wines was evaluated. Among the wines evaluated, Cabernet Sauvignon was the one with the highest activity against all radicals tested. The presence of added copper or iron to wines resulted in a reduced free radical scavenging capacity for all type of wines studied. The formation of redox inactive complexes between polyphenols of wine and transition metals is the possible cause of this reduction in antioxidant activity.
Sigma-Aldrich
Galvinoxyl, free radical