MilliporeSigma
  • Ortho-lithiation of free ferrocenyl alcohols: a new method for the synthesis of planar chiral ferrocene derivatives.
Ortho-lithiation of free ferrocenyl alcohols: a new method for the synthesis of planar chiral ferrocene derivatives.
Chemical communications (Cambridge, England) (2008-07-16)
Bernhard J Ueberbacher, Herfried Griengl, Hansjörg Weber
ABSTRACT
An ortho-metalation method for free ferrocenyl alcohols has been developed, which allows preparation of planar chiral ferrocene derivatives with high yields and diastereoselectivities.
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane