MilliporeSigma
  • Combined physico-chemical treatments based on enterocin AS-48 for inactivation of Gram-negative bacteria in soybean sprouts.
Combined physico-chemical treatments based on enterocin AS-48 for inactivation of Gram-negative bacteria in soybean sprouts.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-06-26)
Antonio Cobo Molinos, Hikmate Abriouel, Rosario Lucas López, Eva Valdivia, Nabil Ben Omar, Antonio Gálvez
ABSTRACT
Enterocin AS-48 was tested for decontamination of soybean sprouts against Gram-negative bacteria. Although treatment with bacteriocin alone had no effect on Salmonella enterica, a synergistic antimicrobial effect was detected at pH 9.0 and in combination with moderate heat treatment. Greatest inactivation was achieved for sprouts heated for 5 min at 65 degrees C in an alkaline (pH 9.0) enterocin AS-48 solution of 25 microg/ml. Bactericidal activity against S. enterica increased greatly when enterocin AS-48 was used in washing solutions in combination with several chemical compounds: EDTA, lactic acid, peracetic acid, polyphosphoric acid, sodium hypochlorite, hexadecylpyridinium chloride, propyl-p-hydroxybenzoate, and hydrocinnamic acid. The combined treatment of enterocin AS-48 and polyphosphoric acid was tested against several other Gram-negative bacteria inoculated on sprouts. The bacteria tested showed great differences in sensitivity to polyphosphoric acid, but synergism with enterocin AS-48 was confirmed in all cases. Combinations of enterocin AS-48 (25 microg/ml) and polyphosphoric acid in a concentration range of 0.1 to 2.0% significantly reduced or inhibited growth of the populations of S. enterica, Escherichia coli O157:H7, Shigella spp., Enterobacter aerogenes, Yersinia enterocolitica, Aeromonas hydrophila and Pseudomonas fluorescens in sprout samples stored at 6 degrees C and 15 degrees C. The combined treatment could therefore be applied to reduce the risks of Gram-negative pathogenic as well as spoilage bacteria on sprouts.
Sigma-Aldrich
Polyphosphoric acid, ~105% H3PO4 basis
Sigma-Aldrich
Polyphosphoric acid, reagent grade, 115% H3PO4 basis