MilliporeSigma
  • Size-controlled synthesis and NMR characterization of mesitylene-vinylsiloxanes stabilized Pt nanoparticles in solution.
Size-controlled synthesis and NMR characterization of mesitylene-vinylsiloxanes stabilized Pt nanoparticles in solution.
Journal of nanoscience and nanotechnology (2008-06-25)
Claudio Evangelisti, Patrizio Raffa, Federica Balzano, Gloria Uccello Barretta, Giovanni Vitulli, Piero Salvadori
ABSTRACT
Pt nanoclusters have been generated by reaction of Pt vapour and mesitylene vapour and the role of the mesitylene/platinum ratio and the Pt particle size has been evaluated, quenching the resulting mesitylene solvated Pt atoms with 1,3-divinyltetramethyldisiloxane (DVS) as additional ligand. The Pt particle sizes have been estimated on the basis of DOSY (Diffusion-Ordered SpectroscopY) analysis and information on their structure features have been obtained by combined use of 2D NMR techniques.
Sigma-Aldrich
Mesitylene, 98%
Supelco
Mesitylene, analytical standard