MilliporeSigma
  • Wastewater quality of natural gas fertilizer factory, Fenchuganj and water quality of Kushiara River at the down stream.
Wastewater quality of natural gas fertilizer factory, Fenchuganj and water quality of Kushiara River at the down stream.
Environmental monitoring and assessment (2008-06-17)
Raquibul Alam, Md Delwar Hossain
ABSTRACT
The physical, chemical and biological characteristics of the wastewater of Natural Gas Fertilizer Factory Ltd. Fenchuganj, Sylhet were determined through extensive laboratory tests in the months of March, July, October and December of the year 2005. Concentration of Suspended Solids was within the range of 445 to 950 mg/L. Bangladesh Industrial Effluent Standards for Suspended Solids is 100 mg/L. Suspended Solids were found above the limit in all the samples. Concentration of Dissolved Solids was found to vary from 576 to 1,456 mg/L. Bangladesh Industrial Effluent Standards, for Dissolved Solids is 2,100 mg/L. Dissolved Solids were found within the limit. Concentration of BOD(5) was found to vary from 4.5 to 8.4 mg/L. Bangladesh Industrial Effluent Standards for BOD(5) is 50 mg/L. For the year 2005, the BOD(5) was found below the limit in all the samples. Dissolved Oxygen of the wastewater was found to be between 2.0 to 3.0 mg/L, which do not satisfy the standard (4.5-8 mg/L). Oil and grease concentration were found in the range of 28 to 68 mg/L, whereas the standard is 10 mg/L for discharge into the inland surface water. In 2005, concentration of Cr(+ 6) was found to vary from 0.01 to 0.156 mg/L in the wastewater. Bangladesh Industrial Effluent Standards, for Cr(+6) is 0.1 mg/L. The concentration of chromium was found above the limit in three samples. Concentration of NO(3) was found to vary from 4.5 to 15.2 mg/L in the wastewater. Bangladesh Industrial Effluent Standards for nitrate is 10 mg/L. The nitrate was found above the limit in three samples. Maximum concentration of ammonia in lagoon 1 was found 1,710 mg/L in month of December. Bangladesh Industrial Effluent Standards for ammonia nitrogen is 100 mg/L. For the year 2005, the ammonia nitrogen was found above the limit in all samples.
Sigma-Aldrich
Chromium(III) nitrate nonahydrate, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Chromium(III) nitrate nonahydrate, 99%
Pricing and availability is not currently available.