MilliporeSigma
Severe infantile hypotonia with ethylmalonic aciduria: case report.
Journal of child neurology (2008-06-10)
Cetin Okuyaz, Fatih Süheyl Ezgü, Gürsel Biberoglu, Massimo Zeviani, Valeria Tiranti, Esat Yilgör
ABSTRACT
An 8-month-old girl was admitted to an outpatient clinic with significant hypotonia and weakness. Organic acid analysis in urine revealed a significant increase in ethylmalonic acid. A deoxyribonucleic analysis revealed the presence of a 625G>A (G-to-A substitution at nucleotide 625) variant short-chain acyl-coenzyme A dehydrogenase gene polymorphism. With the clinical, biochemical and molecular findings, short-chain acyl-coenzyme A dehydrogenase deficiency was suspected. Because 625G>A and 511C>T (C-to-T substitution at nucleotide 511) genetic variations are also present in 14% of the general population, these are considered to be genetic sensitivity variations rather than causing a disease themselves and to result in possible short-chain acyl-coenzyme A dehydrogenase deficiency in the presence of environmental factors such as fever and hunger as well as cellular, biochemical, and other genetic factors. It was stressed that severe infantile hypotonia could also be the only manifestation of ethylmalonic aciduria spectrum disorders.
Sigma-Aldrich
Ethylmalonic acid, 97%