MilliporeSigma
  • The outcome of dimethylglyoxime testing in a sample of cell phones in Denmark.
The outcome of dimethylglyoxime testing in a sample of cell phones in Denmark.
Contact dermatitis (2008-06-10)
Jacob Pontoppidan Thyssen, Jeanne D Johansen, Claus Zachariae, Torkil Menné
ABSTRACT
Nickel dermatitis may be caused by frequent and prolonged use of cell phones. Because little is known about the frequency of nickel release from cell phones, it is difficult to estimate the risk of nickel sensitization and dermatitis among their users. Inspired by a recent case of nickel dermatitis from prolonged cell phone use, the frequency of dimethylglyoxime (DMG)-positive cell phones on the Danish market was investigated. Five major cell phone companies were contacted. Two were visited, and the DMG test was performed on a sample of their products. 5 of 15 (33.3%) phones from company A and 3 of 26 (11.5%) phones from company B showed at least 1 positive reaction. 3 phones had more than 1 positive DMG spots. This study documents that excessive nickel release (i.e. a positive DMG test) is relatively frequent in a sample of cell phones from the Danish market. Prolonged use of cell phones may in some cases fulfil the criteria for items included in the European Union Nickel Directive. We believe that this new cause of nickel dermatitis should be carefully followed and that regulatory steps may be necessary.
Sigma-Aldrich
Dimethylglyoxime, ACS reagent, ≥99%
Sigma-Aldrich
Dimethylglyoxime, ≥97.0% (TLC)
Sigma-Aldrich
Dimethylglyoxime, puriss. p.a., ACS reagent, for the detection of Ni, ≥99.0% (TLC)