MilliporeSigma
  • Effect of decabromodiphenyl ether and antimony trioxide on controlled pyrolysis of high-impact polystyrene mixed with polyolefins.
Effect of decabromodiphenyl ether and antimony trioxide on controlled pyrolysis of high-impact polystyrene mixed with polyolefins.
Chemosphere (2008-05-24)
Nona Merry M Mitan, Thallada Bhaskar, William J Hall, Akinori Muto, Paul T Williams, Yusaku Sakata
ABSTRACT
The controlled pyrolysis of polyethylene/polypropylene/polystyrene mixed with brominated high-impact polystyrene containing decabromodiphenyl ether as a brominated flame-retardant with antimony trioxide as a synergist was performed. The effect of decabromodiphenyl ether and antimony trioxide on the formation of its congeners and their effect on distribution of pyrolysis products were investigated. The controlled pyrolysis significantly affected the decomposition behavior and the formation of products. Analysis with gas chromatograph with electron capture detector confirmed that the bromine content was rich in step 1 (oil 1) liquid products leaving less bromine content in the step 2 (oil 2) liquid products. In the presence of antimony containing samples, the major portion of bromine was observed in the form of antimony bromide and no flame-retardant species were found in oil 1. In the presence of synergist, the step 1 and step 2 oils contain both light and heavy compounds. In the absence of synergist, the heavy compounds in step 1 oil and light compounds in step 2 oils were observed. The presence of antimony bromide was confirmed in the step 1 oils but not in step 2 oils.
Sigma-Aldrich
Antimony(III) oxide, nanopowder, <250 nm particle size (TEM), ≥99.9% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, powder, 5 μm, ReagentPlus®, 99%