MilliporeSigma
  • Toward autonomously operating molecular machines driven by transition-metal catalyst.
Toward autonomously operating molecular machines driven by transition-metal catalyst.
Molecular bioSystems (2008-05-22)
Kenichi Tanaka, Kazushi Kinbara
ABSTRACT
A Vaska-type complex carrying a ferrocene-appended diphosphine ligand was developed as a prototype of autonomous molecular machines that is able to undergo continuous scissoring motion in the presence of diphenylphosphoryl azide and aldehyde.
Sigma-Aldrich
Diphenyl phosphoryl azide, 97%