MilliporeSigma
  • Aqueous solubility and Daphnia magna chronic toxicity of di(2-ethylhexyl) adipate.
Aqueous solubility and Daphnia magna chronic toxicity of di(2-ethylhexyl) adipate.
Bulletin of environmental contamination and toxicology (2008-05-17)
K A Robillard, D L Dufresne, J W Gorsuch, W A Stubblefield, C A Staples, T F Parkerton
ABSTRACT
A water solubility of 5.5 (+/-0.22) microg/L for di(2-ethylhexyl) adipate (DEHA) was measured using the slow-stir method. This value is consistent with computer estimations and over two orders of magnitude lower than that previously determined using the shake-flask method. We performed a 21-day chronic Daphnia magna limit test at an average exposure of 4.4 microg/L in laboratory diluent water to avoid insoluble test material and avoid physical entrapment. One hundred percent of the DEHA-treated organisms survived compared to 90% survival in both the controls and solvent controls. Mean neonate reproduction was 152, 137, and 148 and mean dry weight per surviving female was 0.804, 0.779, and 0.742 mg in the DEHA treatment, control, and solvent control, respectively. No adverse effects were observed.
Product Description
Supelco
Bis(2-ethylhexyl) adipate, 99%
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥97.0% (GC)
Supelco
Bis(2-ethylhexyl) adipate, Selectophore, ≥99.0%
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥99%
Supelco
Di(2-ethylhexyl)adipate solution, certified reference material, 2000 μg/mL in methanol