MilliporeSigma
  • DFT calculation of 1J(119Sn,13C) and 2J(119Sn,1H) coupling constants in di- and trimethyltin(IV) compounds.
DFT calculation of 1J(119Sn,13C) and 2J(119Sn,1H) coupling constants in di- and trimethyltin(IV) compounds.
Inorganic chemistry (2008-05-08)
Girolamo Casella, Francesco Ferrante, Giacomo Saielli
ABSTRACT
We have tested several computational protocols, at the nonrelativistic DFT level of theory, for the calculation of 1J(119Sn, 13C) and 2J(119Sn, 1H) spin-spin coupling constants in di- and trimethyltin(IV) derivatives with various ligands. Quite a good agreement with experimental data has been found with several hybrid functionals and a double-zeta basis set for a set of molecules comprising tetra-, penta-, and hexa-coordinated tin(IV). Then, some of the protocols have been applied to the calculation of the 2J(119Sn, 1H) of the aquodimethyltin(IV) ion and dimethyltin(IV) complex with D-ribonic acid and to the calculation of 1J(119Sn, 13C) and 2J(119Sn, 1H) of the dimethyltin(IV)-glycylglycine and glycylhistidine complexes in water solutions. Solvent effects have been considered in these cases by including explicit water molecules and/or the solvent reaction field, resulting in a good agreement with experimental data. The proposed protocols constitute a helpful tool for the structural determination of di- and triorganotin(IV) derivatives.
Sigma-Aldrich
Gly-His hydrochloride hydrate, ≥99.0% (calc. based on dry substance, AT)
Sigma-Aldrich
Gly-His