MilliporeSigma
  • Speciation of Cr(III) and Cr(VI) in environmental samples determined by selective separation and preconcentration on silica gel chemically modified with niobium(V) oxide.
Speciation of Cr(III) and Cr(VI) in environmental samples determined by selective separation and preconcentration on silica gel chemically modified with niobium(V) oxide.
Journal of hazardous materials (2008-05-06)
Edmar Martendal, Heloisa França Maltez, Eduardo Carasek
ABSTRACT
In this study a new method for chromium speciation in water using solid phase extraction coupled to a flow injection system and flame atomic absorption spectrometry was developed. The adsorption behavior of Cr(III) and Cr(VI) on Nb2O5-SiO2 allowed the selective separation of Cr(III) from Cr(VI) in the pH range of 6-9. Thus, a method for Cr(III) preconcentration and extraction using Nb2O5-SiO2 was developed. Total chromium was determined after the reduction of Cr(VI) to Cr(III) using sodium sulfite in acidic medium. The operational variables of the preconcentration and reduction procedures were optimized through full factorial and Doehlert designs. The limit of detection for Cr(III) was 0.34microgL(-1) and the precision was below 4.6%. Results for recovery tests using different environmental samples were between 90 and 105%. Certified reference materials (NIST 1640 and NIST 1643e) were analyzed in order to check the accuracy of the proposed method, and the results were in agreement with the certified values.
Sigma-Aldrich
Niobium(V) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Niobium(V) oxide, 99.99% trace metals basis