MilliporeSigma
  • Measurement of residual solvents in a drug substance by a purge-and-trap method.
Measurement of residual solvents in a drug substance by a purge-and-trap method.
Journal of pharmaceutical and biomedical analysis (2008-04-26)
Miklós Lakatos
ABSTRACT
The purge-and-trap (P&T) gas extraction method combined with gas chromatography was studied for its suitability for quantitative residual solvents determination in a water-soluble active pharmaceutical ingredient (API). Some analytical method performance characteristics were investigated, namely, the repeatability, the accuracy and the detection limit of determination. The results show that the P&T technique is--as expected--more sensitive than the static headspace, thus it can be used for the determination of residual solvents pertaining to the ICH Class 1 group. It was found that it could be an alternative sample preparation method besides the static headspace (HS) method.
Product Description
Supelco
1,1-Dichloroethane, analytical standard, contains 100 ppm hydroquinone as stabilizer
Supelco
1,1-Dichloroethane solution, certified reference material, 5000 μg/mL in methanol