MilliporeSigma
  • Molecular fibers and wires in solid-state and solution self-assemblies of cyclodextrin [2]rotaxanes.
Molecular fibers and wires in solid-state and solution self-assemblies of cyclodextrin [2]rotaxanes.
Organic letters (2008-04-16)
Subashani Maniam, Marta M Cieslinski, Stephen F Lincoln, Hideki Onagi, Peter J Steel, Anthony C Willis, Christopher J Easton
ABSTRACT
Cyclodextrin [2]rotaxanes have been prepared by coupling dimethylanilines with dicarboxylic acids using DMT-MM, in aqueous solutions of alpha-cyclodextrin, and the example illustrated shows unusual fluorescence emission and other spectroscopic behavior characteristic of the formation of molecular wires in solution, similar to the fibers observed in the solid state.
Sigma-Aldrich
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
Sigma-Aldrich
N,N-Dimethylaniline, ReagentPlus®, 99%