MilliporeSigma
  • Electrokinetic probes for single-step screening of polyol stereoisomers: the virtues of ternary boronate ester complex formation.
Electrokinetic probes for single-step screening of polyol stereoisomers: the virtues of ternary boronate ester complex formation.
Chemical communications (Cambridge, England) (2008-04-11)
Claire Kaiser, Giselle Segui-Lines, Jason C D'Amaral, Adam S Ptolemy, Philip Britz-McKibbin
ABSTRACT
Electrokinetic probes based on the differential migration of ternary boronate ester complexes permit the selective analysis of micromolar levels of UV-transparent polyol stereoisomers in urine samples via dynamic complexation-capillary electrophoresis that is applicable to single-step screening of in-born errors of sugar metabolism, such as galactosemia.
Sigma-Aldrich
3-Nitrophenylboronic acid, ≥97%