MilliporeSigma
  • A convenient route to enantiopure 3-aryl-2,3-diaminopropanoic acids by diastereoselective Mannich reaction of camphor-based tricyclic iminolactone with imines.
A convenient route to enantiopure 3-aryl-2,3-diaminopropanoic acids by diastereoselective Mannich reaction of camphor-based tricyclic iminolactone with imines.
The Journal of organic chemistry (2008-03-28)
Huan-Huan Zhang, Xiu-Qin Hu, Xiao Wang, Yong-Chun Luo, Peng-Fei Xu
ABSTRACT
A novel and convenient route to the asymmetric synthesis of 2,3-diamino acids via Mannich reaction of iminolactones 1a and 1b with N-protected imines has been achieved in good yields (up to 95%) and high diastereoselectivity (dr: >99:1). Hydrolysis of the Mannich adducts under acidic conditions furnished the desired 3-aryl-2,3-diaminopropanoic acids in good yields (up to 85%) with excellent enantiomeric excesses (99% ee).
Sigma-Aldrich
DL-2,3-Diaminopropionic acid monohydrochloride, 98%