MilliporeSigma
  • Reactivity of N-(omega-haloalkyl)-beta-lactams with regard to lithium aluminium hydride: novel synthesis of 1-(1-aryl-3-hydroxypropyl)aziridines and 3-aryl-3-(N-propylamino)propan-1-ols.
Reactivity of N-(omega-haloalkyl)-beta-lactams with regard to lithium aluminium hydride: novel synthesis of 1-(1-aryl-3-hydroxypropyl)aziridines and 3-aryl-3-(N-propylamino)propan-1-ols.
Organic & biomolecular chemistry (2008-03-26)
Matthias D'hooghe, Stijn Dekeukeleire, Norbert De Kimpe
ABSTRACT
The reactivity of 4-aryl-1-(2-chloroethyl)azetidin-2-ones and 4-aryl-1-(3-bromopropyl)azetidin-2-ones with regard to lithium aluminium hydride has been evaluated for the first time. 4-Aryl-1-(2-chloroethyl)azetidin-2-ones were transformed into novel 1-(1-aryl-3-hydroxypropyl)aziridines through an unprecedented conversion of beta-lactams into 2,3-unsubstituted aziridine derivatives. Unexpectedly, 4-aryl-1-(3-bromopropyl)azetidin-2-ones underwent dehalogenation towards 3-aryl-3-(N-propylamino)propan-1-ols upon treatment with LiAlH(4). 1-(1-Aryl-3-hydroxypropyl)aziridines were further elaborated by means of ring opening reactions using benzyl bromide in acetonitrile towards 3-aryl-3-[N-benzyl-N-(2-bromoethyl)amino]propan-1-ols and using aluminium(iii) chloride in diethyl ether, affording 3-aryl-3-[N-(2-chloroethyl)amino]propan-1-ols.
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF