MilliporeSigma
  • In vitro assessment of mitochondrial dysfunction and cytotoxicity of nefazodone, trazodone, and buspirone.
In vitro assessment of mitochondrial dysfunction and cytotoxicity of nefazodone, trazodone, and buspirone.
Toxicological sciences : an official journal of the Society of Toxicology (2008-03-18)
James A Dykens, Joseph D Jamieson, Lisa D Marroquin, Sashi Nadanaciva, Jinghai J Xu, Margaret C Dunn, Arthur R Smith, Yvonne Will
ABSTRACT
Mitochondrial toxicity is increasingly implicated in a host of drug-induced organ toxicities, including hepatotoxicity. Nefazodone was withdrawn from the U.S. market in 2004 due to hepatotoxicity. Accordingly, we evaluated nefazodone, another triazolopyridine trazodone, plus the azaspirodecanedione buspirone, for cytotoxicity and effects on mitochondrial function. In accord with its clinical disposition, nefazodone was the most toxic compound of the three, trazodone had relatively modest effects, whereas buspirone showed the least toxicity. Nefazodone profoundly inhibited mitochondrial respiration in isolated rat liver mitochondria and in intact HepG2 cells where this was accompanied by simultaneous acceleration of glycolysis. Using immunocaptured oxidative phosphorylation (OXPHOS) complexes, we identified Complex 1, and to a lesser amount Complex IV, as the targets of nefazodone toxicity. No inhibition was found for trazodone, and buspirone showed 3.4-fold less inhibition of OXPHOS Complex 1 than nefazodone. In human hepatocytes that express cytochrome P450, isoform 3A4, after 24 h exposure, nefazodone and trazodone collapsed mitochondrial membrane potential, and imposed oxidative stress, as detected via glutathione depletion, leading to cell death. Our results suggest that the mitochondrial impairment imposed by nefazodone is profound and likely contributes to its hepatotoxicity, especially in patients cotreated with other drugs with mitochondrial liabilities.
Sigma-Aldrich
Nefazodone hydrochloride, ≥98% (HPLC), solid